COM448619185 Combi-Blocks, Inc.

1-(2-Bromo-4-nitrophenyl)-4-hydroxypiperidine

  • MDL number MFCD11039803
  • Cas Number : 1072944-61-6
  • Molecular Weight : 301.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2132
Availability details for
Quantity
  • Availability
  • Add to Cart