COM448619190 Combi-Blocks, Inc.

2,3-Dimethoxy-6-nitropyridine

  • MDL number MFCD11044329
  • Cas Number : 79491-44-4
  • Molecular Weight : 184.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2138

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2138
Availability details for
Quantity
  • Availability
  • Add to Cart