COM448619208 Combi-Blocks, Inc.

3,6-Dibromo-7-methylimidazo[1,2-a]pyridine, HCl

  • MDL number MFCD11855860
  • Cas Number : 1072944-65-0
  • Molecular Weight : 326.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2158

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2158
Availability details for
Quantity
  • Availability
  • Add to Cart