COM448619213 Combi-Blocks, Inc.

3-Bromo-6-fluoro-4-hydroxy-2-trifluoromethylquinoline

  • MDL number MFCD11504911
  • Cas Number : 1072944-66-1
  • Molecular Weight : 310.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2163
Availability details for
Quantity
  • Availability
  • Add to Cart