COM448619219 Combi-Blocks, Inc.

4,7-Dichloro-8-methyl-2-(trifluoromethyl)quinoline

  • MDL number MFCD08448261
  • Cas Number : 887350-78-9
  • Molecular Weight : 280.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2172

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2172
Availability details for
Quantity
  • Availability
  • Add to Cart