COM448619226 Combi-Blocks, Inc.

4-Bromo-3-phenyl-1(2)H-pyrazole

  • MDL number MFCD11855861
  • Cas Number : 13808-65-6
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-5-phenyl-1H-pyrazole
Inchi Code 1S/C9H7BrN2/c10-8-6-11-12-9(8)7-4-2-1-3-5-7/h1-6H,(H,11,12)
Inchi Key ZJFUSLGMGSYTRY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2206

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2206
Availability details for
Quantity
  • Availability
  • Add to Cart