COM448619230 Combi-Blocks, Inc.

Ethyl 5-amino-1-(4-chlorophenyl)-4-cyanopyrazole-3-carboxylate

  • MDL number MFCD09835737
  • Cas Number : 908584-68-9
  • Molecular Weight : 290.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2211

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2211
Availability details for
Quantity
  • Availability
  • Add to Cart