COM448619239 Combi-Blocks, Inc.

5-Amino-8-fluoroquinoline

  • MDL number MFCD09042730
  • Cas Number : 175229-87-5
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2225

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2225
Availability details for
Quantity
  • Availability
  • Add to Cart