COM448619241 Combi-Blocks, Inc.

5-p-Tolyl-1H-pyrazole

  • MDL number MFCD08446303
  • Cas Number : 59843-75-3
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2227
Availability details for
Quantity
  • Availability
  • Add to Cart