COM448619253 Combi-Blocks, Inc.

4-Bromo-3-p-tolyl-1(2)H-pyrazole

  • MDL number MFCD11855866
  • Cas Number : 1116093-45-8
  • Molecular Weight : 237.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2258

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2258
Availability details for
Quantity
  • Availability
  • Add to Cart