COM448619259 Combi-Blocks, Inc.

Methyl 5,7-dichloro-4-hydroxyquinoline-2-carboxylate

  • MDL number MFCD11855869
  • Cas Number : 130613-19-3
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2266
Availability details for
Quantity
  • Availability
  • Add to Cart