COM448619260 Combi-Blocks, Inc.

Methyl 8-bromo-5-fluoro-4-hydroxyquinoline-2-carboxylate

  • MDL number MFCD11855870
  • Cas Number : 1133115-48-6
  • Molecular Weight : 300.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2267

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2267
Availability details for
Quantity
  • Availability
  • Add to Cart