COM448619266 Combi-Blocks, Inc.

Methyl 2-amino-4-(4-chlorophenyl)pyrimidine-5-carboxylate

  • MDL number MFCD11855874
  • Cas Number : 1133115-56-6
  • Molecular Weight : 263.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2276

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2276
Availability details for
Quantity
  • Availability
  • Add to Cart