COM448619277 Combi-Blocks, Inc.

4-Bromo-5-(3-chlorophenyl)pyrazole

  • MDL number MFCD11855879
  • Cas Number : 149739-38-8
  • Molecular Weight : 257.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2291

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2291
Availability details for
Quantity
  • Availability
  • Add to Cart