COM448619279 Combi-Blocks, Inc.

Ethyl 5-amino-1-(4-bromophenyl)-4-cyanopyrazole-3-carboxylate

  • MDL number MFCD09835740
  • Cas Number : 96723-27-2
  • Molecular Weight : 335.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2294

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2294
Availability details for
Quantity
  • Availability
  • Add to Cart