COM448619324 Combi-Blocks, Inc.

2-bromo-5-methoxy-1,3-benzothiazole

  • MDL number MFCD09749263
  • Cas Number : 214337-39-0
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2369

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2369
Availability details for
Quantity
  • Availability
  • Add to Cart