COM448619326 Combi-Blocks, Inc.

2-bromo-5-chloro-1,3-benzothiazole

  • MDL number MFCD09749223
  • Cas Number : 2941-56-2
  • Molecular Weight : 248.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2372

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2372
Availability details for
Quantity
  • Availability
  • Add to Cart