COM448619500 Combi-Blocks, Inc.

1-(2-Bromo-4-nitrophenyl)piperidine

  • MDL number MFCD11039794
  • Cas Number : 1260658-32-9
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2685

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2685
Availability details for
Quantity
  • Availability
  • Add to Cart