COM448619609 Combi-Blocks, Inc.

N-(5-Methylpyrazin-2-yl)acetamide

  • MDL number MFCD24558363
  • Cas Number : 5521-59-5
  • Molecular Weight : 151.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2883

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2883
Availability details for
Quantity
  • Availability
  • Add to Cart