COM448619650 Combi-Blocks, Inc.

1-bromo-2-iodo-5-methoxy-3-nitrobenzene

  • MDL number MFCD18379738
  • Cas Number : 1263376-99-3
  • Molecular Weight : 357.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2943

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2943
Availability details for
Quantity
  • Availability
  • Add to Cart