COM448619768 Combi-Blocks, Inc.

1,2-dichloro-4-iodo-5-nitrobenzene

  • MDL number MFCD26958724
  • Cas Number : 1774898-47-3
  • Molecular Weight : 317.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3152

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3152
Availability details for
Quantity
  • Availability
  • Add to Cart