COM448619853 Combi-Blocks, Inc.

1-methyl-2-nitro-4-(trifluoromethoxy)benzene

  • MDL number MFCD09954767
  • Cas Number : 70692-45-4
  • Molecular Weight : 221.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3282

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3282
Availability details for
Quantity
  • Availability
  • Add to Cart