COM448619951 Combi-Blocks, Inc.

3-Bromo-5,6-dimethoxypyridin-2-amine

  • MDL number MFCD28122672
  • Cas Number : 1820665-96-0
  • Molecular Weight : 233.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3459

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3459
Availability details for
Quantity
  • Availability
  • Add to Cart