COM448620142 Combi-Blocks, Inc.

2-Acetamido-5-methylthiazole

  • MDL number MFCD18801061
  • Cas Number : 61996-32-5
  • Molecular Weight : 156.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3777
Availability details for
Quantity
  • Availability
  • Add to Cart