COM448620335 Combi-Blocks, Inc.

Ethyl 5-tert-butylpyrazole-3-carboxylate

  • MDL number MFCD00173782
  • Cas Number : 83405-70-3
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4029
Availability details for
Quantity
  • Availability
  • Add to Cart