COM448620393 Combi-Blocks, Inc.

2,3-Difluorobenzylamine HCl

  • MDL number MFCD13185548
  • Cas Number : 1214350-73-8
  • Molecular Weight : 179.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4099
Availability details for
Quantity
  • Availability
  • Add to Cart