COM448620430 Combi-Blocks, Inc.

1-Fluoro-2-isopropoxy-4-nitrobenzene

  • MDL number MFCD23114386
  • Cas Number : 1229647-66-8
  • Molecular Weight : 199.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4145
Availability details for
Quantity
  • Availability
  • Add to Cart