COM448620435 Combi-Blocks, Inc.

2-(3-Chloropropyl)isoindole-1,3-dione

  • MDL number MFCD00158628
  • Cas Number : 42251-84-3
  • Molecular Weight : 223.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4156

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4156
Availability details for
Quantity
  • Availability
  • Add to Cart