COM448620437 Combi-Blocks, Inc.

5-Bromo-2-fluoro-4-methylaniline HCl

  • MDL number MFCD27981306
  • Cas Number : 1820619-16-6
  • Molecular Weight : 240.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4160

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4160
Availability details for
Quantity
  • Availability
  • Add to Cart