COM448620447 Combi-Blocks, Inc.

1-Benzoyl-3-(2-hydroxy-5-methylphenyl)thiourea

  • MDL number MFCD02631941
  • Cas Number : 429642-03-5
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4171

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4171
Availability details for
Quantity
  • Availability
  • Add to Cart