COM448620448 Combi-Blocks, Inc.

1-Ethyl-3-(trifluoromethyl)pyrazole-4-carbaldehyde

  • MDL number MFCD25949488
  • Cas Number : 1443279-72-8
  • Molecular Weight : 192.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4172

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4172
Availability details for
Quantity
  • Availability
  • Add to Cart