COM448620461 Combi-Blocks, Inc.

Ethyl 2-(4-fluorophenyl)-2-methylpropanoate

  • MDL number MFCD16748922
  • Cas Number : 1035261-07-4
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4188

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4188
Availability details for
Quantity
  • Availability
  • Add to Cart