COM448620464 Combi-Blocks, Inc.

3-(Dimethoxymethyl)-2-fluorobenzaldehyde

  • MDL number MFCD28014029
  • Cas Number : 1820705-34-7
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4191

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4191
Availability details for
Quantity
  • Availability
  • Add to Cart