COM448620465 Combi-Blocks, Inc.

2-(Benzyloxy)aniline hydrochloride

  • MDL number MFCD27960265
  • Cas Number : 857594-21-9
  • Molecular Weight : 235.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4192

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4192
Availability details for
Quantity
  • Availability
  • Add to Cart