COM448620467 Combi-Blocks, Inc.

4-Bromo-1-(methoxymethyl)pyrazole

  • MDL number MFCD18324047
  • Cas Number : 1071200-42-4
  • Molecular Weight : 191.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4194

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4194
Availability details for
Quantity
  • Availability
  • Add to Cart