COM448620468 Combi-Blocks, Inc.

1-(Methoxymethyl)pyrrole-2-carboxylic acid

  • MDL number MFCD18810222
  • Cas Number : 172805-62-8
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4195

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4195
Availability details for
Quantity
  • Availability
  • Add to Cart