COM448620470 Combi-Blocks, Inc.

2,2-Diethyl 7-phenyl-6,8-dioxaspiro[3.5]nonane-2,2-dicarboxylate

  • MDL number MFCD00101441
  • Cas Number : 142733-60-6
  • Molecular Weight : 348.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4197

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4197
Availability details for
Quantity
  • Availability
  • Add to Cart