COM448620471 Combi-Blocks, Inc.

N-[2-(2-bromophenyl)ethyl]-2,2,2-trifluoroacetamide

  • MDL number MFCD28096633
  • Cas Number : 1057246-44-2
  • Molecular Weight : 296.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4200

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4200
Availability details for
Quantity
  • Availability
  • Add to Cart