COM448620476 Combi-Blocks, Inc.

2-[(3-Fluorophenyl)carbonyl]piperidine hydrochloride

  • MDL number MFCD28122673
  • Cas Number : 1820734-90-4
  • Molecular Weight : 243.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4205

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4205
Availability details for
Quantity
  • Availability
  • Add to Cart