COM448620478 Combi-Blocks, Inc.

tert-Butyl 4-(4-bromo-2-chlorophenyl)piperazine-1-carboxylate

  • MDL number MFCD25973037
  • Cas Number : 1429891-40-6
  • Molecular Weight : 375.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4207

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4207
Availability details for
Quantity
  • Availability
  • Add to Cart