COM448620480 Combi-Blocks, Inc.

N-(3-Bromophenethyl)-2,2-difluoroacetamide

  • MDL number MFCD26404250
  • Cas Number : 1010422-41-9
  • Molecular Weight : 278.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4209

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4209
Availability details for
Quantity
  • Availability
  • Add to Cart