COM448620484 Combi-Blocks, Inc.

Methyl 2-amino-4-(4-bromophenyl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD28101639
  • Cas Number : 1284363-17-2
  • Molecular Weight : 313.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4214

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4214
Availability details for
Quantity
  • Availability
  • Add to Cart