COM448620485 Combi-Blocks, Inc.

4-Bromo-1-[(4-methylbenzene)sulfonyl]pyrazole

  • MDL number MFCD06080375
  • Cas Number : 116228-41-2
  • Molecular Weight : 301.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4215

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4215
Availability details for
Quantity
  • Availability
  • Add to Cart