COM448620493 Combi-Blocks, Inc.

1-Bromo-5-chloro-2-fluoro-3-nitrobenzene

  • MDL number MFCD28143933
  • Cas Number : 1679357-80-2
  • Molecular Weight : 254.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4227

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4227
Availability details for
Quantity
  • Availability
  • Add to Cart