COM448620495 Combi-Blocks, Inc.

3-Bromo-5-chloro-2-fluoroaniline hydrochloride

  • MDL number MFCD22573644
  • Cas Number : 1384265-18-2
  • Molecular Weight : 260.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4229

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4229
Availability details for
Quantity
  • Availability
  • Add to Cart