COM448620500 Combi-Blocks, Inc.

(2-Bromo-4-chloro-6-nitrophenyl)acetic acid

  • MDL number MFCD11845449
  • Cas Number : 1082040-47-8
  • Molecular Weight : 294.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4235

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4235
Availability details for
Quantity
  • Availability
  • Add to Cart