COM448620507 Combi-Blocks, Inc.

2-(4-BOC-Piperazino)methyl-4-bromothiophene

  • MDL number MFCD28369582
  • Cas Number : 1820603-98-2
  • Molecular Weight : 361.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4242

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4242
Availability details for
Quantity
  • Availability
  • Add to Cart