COM448620520 Combi-Blocks, Inc.

2,3-Diaminofluorobenzene hydrochloride

  • MDL number MFCD22689205
  • Cas Number : 1414959-15-1
  • Molecular Weight : 162.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4261

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4261
Availability details for
Quantity
  • Availability
  • Add to Cart