COM448620527 Combi-Blocks, Inc.

Benzyl N-[1-(2-fluorophenyl)cyclopropyl]carbamate

  • MDL number MFCD28397268
  • Cas Number : 1820648-90-5
  • Molecular Weight : 285.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4270
Availability details for
Quantity
  • Availability
  • Add to Cart