COM448620615 Combi-Blocks, Inc.

2-Chloro-4-isopropylpyrimidine

  • MDL number MFCD09475913
  • Cas Number : 941294-36-6
  • Molecular Weight : 156.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6113

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-6113
Availability details for
Quantity
  • Availability
  • Add to Cart